missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™488-conjugated NEUROD1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66691-100UL
This item is not returnable.
View return policy
Description
The NEUROD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NEUROD1. This antibody recognizes human antigen. The NEUROD1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.Specifications
| NEUROD1 | |
| Monoclonal | |
| 1667 μg/mL | |
| BETA2; BHF-1; NEUROD; bHLHa3 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human | |
| 40-50 kDa | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 3E10F1 | |
| CoraLite 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 4760 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction