missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™488-conjugated NEUROD1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | NEUROD1 |
|---|---|
| Clone | 3E10F1 |
| Concentration | 1667 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237712
|
Proteintech
CL488-66691-20UL |
20 μL |
140.00€ 129.00€ / 20µL Save 11.00€ 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234733
|
Proteintech
CL488-66691-100UL |
100 μL |
450.00€ 349.00€ / 100µL Save 101.00€ 22% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NEUROD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NEUROD1. This antibody recognizes human antigen. The NEUROD1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.Specifications
| NEUROD1 | |
| 1667 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 4760 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3E10F1 | |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite 488 | |
| Mouse | |
| Human | |
| BETA2; BHF-1; NEUROD; bHLHa3 | |
| IgG1 | |
| Protein G purification | |
| Conjugated Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title