missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™488-conjugated NEUROD1 Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL488-66691-20UL

 View more versions of this product

Product Code. 30237712

  • ONLINE EXCLUSIVE PRICE
    140.00€
    129.00€ / 20µL
    Save 11.00€
    7% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The NEUROD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NEUROD1. This antibody recognizes human antigen. The NEUROD1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specifications

Specifications

NEUROD1
Monoclonal
1667 μg/mL
BETA2; BHF-1; NEUROD; bHLHa3
Recombinant Protein
20 μL
Primary
Human
40-50 kDa
Liquid
Flow Cytometry (Intracellular Staining)
3E10F1
CoraLite 488
Mouse
Protein G purification
RUO
4760
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Conjugated Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.