Learn More
Eppendorf™ Concentrator Plus Rotors
Shop All Eppendorf Products
Click to view available options
Capacity:
16 x 6.5/10 mL
18 x 10 mL
2 x 96 x 0.2 mL
24 x 6 mL
36 x 1.5 mL
6 x 15/50 mL
8 x 15 mL
8 x 16 mL
8 x 25 mL
Description:
Rotor A-2-VC
Rotor F-35-6-30
Rotor F-35-8-24
Rotor F-40-18-19
Rotor F-40-36-12
Rotor F-45-16-20
Rotor F-45-24-12
Rotor F-45-8-17
Rotor F-50-8-16
Rotor F-50-8-18
Unit Size:
Each
Description
- Advanced heating technology provides best treatment for your sample which assures quick, efficient and gentle vacuum concentration of DNA/RNA, nucleotides, proteins and other liquid or wet samples. Our new coated lid provides superior chemical resistance against aggressive acids and organic solvents (e.g. TFA, DMSO).
- The Concentrator plus is available with three configurations to meet the customers' lab specific needs:
- Basic Concentrator for connecting to an existing vacuum pump
- Complete system with an integrated solvent-resistant, maintenance and oil-free PTFE pump, and condensation trap
- Complete system with vacuum port adds extra capability. It allows for using the integrated vacuum pump separately – without having to disconnect it from the system
- Choice of 3 operation modes (vacufuge, centrifuge, and desiccator) for a variety of application needs
- Choice of three application modes (aqueous, alcohol or high vapor pressure) to correspond with sample solvent—reduction of processing time up to 20%
- Choice of 4 heating levels (ambient, 30C, 45°C, 60°C) allows safe and efficient concentration of various samples
- Brake function can optionally be turned off for use with delicate samples
Specifications
Specifications
| Description | Rotor F-40-36-12 |
| Type | Concentrator Plus Rotor |
| Capacity | 36 x 1.5 mL |
| Max. Tube Length | 32 mm |
| Max. Tube Diameter | 12 mm |
| For Use With (Equipment) | Concentrator Plus Centrifuge |