missing translation for 'onlineSavingsMsg'
Learn More

Complement Factor H Mouse anti-Human, Clone: C18/3, Novus Biologicals™

Mouse Monoclonal Antibody

222.00€ - 520.00€

Specifications

Antigen Complement Factor H
Clone C18/3
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Applications Western Blot, ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18131645
View Documents
Novus Biologicals
NBP2-23542-100UG
100 μg
520.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18187002
View Documents
Novus Biologicals
NBP2-23542-0.025MG
0.025 mg
222.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Complement Factor H Monoclonal specifically detects Complement Factor H in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

Complement Factor H
1 mg/mL
Western Blot, ELISA
Unconjugated
Mouse
Human
adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246
CFH
IgG1 κ
Protein A or G purified
Clone C18/3 is specific for complement factor H in human serum and plasma
C18/3
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Monoclonal
Purified
RUO
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
3075
Human Factor H
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.