missing translation for 'onlineSavingsMsg'
Learn More

Complement Factor H Mouse anti-Human, Clone: C18/3, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-23542-100UG

 View more versions of this product

Product Code. 18131645

  • 520.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Complement Factor H Monoclonal specifically detects Complement Factor H in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Complement Factor H
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246
Mouse
Protein A or G purified
RUO
3075
Human
Purified
Western Blot, ELISA
C18/3
Unconjugated
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
CFH
Human Factor H
100 μg
Primary
Clone C18/3 is specific for complement factor H in human serum and plasma
Store at 4C. Do not freeze.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only