missing translation for 'onlineSavingsMsg'
Learn More

CLEC10A/CD301 Antibody (CL15513) [Unconjugated], Novus Biologicals™

Mouse Monoclonal Antibody

422.00€ - 605.00€

Specifications

Antigen CLEC10A/CD301
Clone CL15513
Concentration 1 mg/mL
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30279749
View Documents Promotion Details
Novus Biologicals
NBP3-43672-100ul
100 μL
605.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30279395
View Documents Promotion Details
Novus Biologicals
NBP3-43672-25ul
25 μL
422.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CLEC10A/CD301 Monoclonal antibody specifically detects CLEC10A/CD301 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

CLEC10A/CD301
1 mg/mL
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Immunology
PBS, pH 7.2, 40% glycerol
10462
IgG2b
Protein A purified
CL15513
Western Blot 1 μg/mL, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Monoclonal
Purified
RUO
Human
CD301 antigen, CD301C-type lectin domain family 10 member A, CLECSF13, CLECSF14macrophage C-type lectin, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 14 (macrophage-derived), C-type lectin domain family 10, member A, C-type lectin superfamily member 14, HML2, HMLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 13 (macrophage-derived), Macrophage lectin 2, macrophage lectin 2 (calcium dependent), MGL
This antibody was generated using a synthetic peptide of Q8IUN9, with the exact immunogen sequence remaining proprietary.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.