missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC10A/CD301 Antibody (CL15513) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43672-100ul
This item is not returnable.
View return policy
Description
CLEC10A/CD301 Monoclonal antibody specifically detects CLEC10A/CD301 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| CLEC10A/CD301 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| CD301 antigen, CD301C-type lectin domain family 10 member A, CLECSF13, CLECSF14macrophage C-type lectin, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 14 (macrophage-derived), C-type lectin domain family 10, member A, C-type lectin superfamily member 14, HML2, HMLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 13 (macrophage-derived), Macrophage lectin 2, macrophage lectin 2 (calcium dependent), MGL | |
| This antibody was generated using a synthetic peptide of Q8IUN9, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Immunology | |
| 10462 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL15513 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction