Learn More
Fisherbrand™ Clear Standard Wide Mouth Glass Bottles without Caps
GREENER_CHOICE
Product Code. 16104627 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
120 mL
240 mL
950 mL
2000 mL
3840 mL
Unit Size:
Pack of 12
Pack of 24
Pack of 4
Pack of 6
Product Code. 16104627
Only null left
Add to basket
Add to basket
Clear, Type III soda-lime glass ideal for general, small to large volume storage
  • Features a wide mouth opening for easy transfer of contents
  • Applications include pharmaceutical, chemical, environmental, and food packaging and storing
  • Manufactured from clear flint glass
Specifications
Capacity (Metric) 950 mL
Capacity (English) 32 oz.
Color Clear
Material Glass (Type 3)
Quantity 12
Neck Finish 89-400
Closure Included No
Mouth Wide
Product Type Standard Wide Mouth Bottle
Closure Size 89-400 mm