missing translation for 'onlineSavingsMsg'
Learn More

cis-Atovaquone (Racemic), TRC

Product Code. 30008343
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008343 10 mg 10mg
30003236 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008343 Supplier Toronto Research Chemicals Supplier No. TRCA79345010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137732-39-9
Molecular Formula C22 H19 Cl O3
Molecular Weight (g/mol) 366.84
Synonym cis-2-[4-(4-chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthoquinone, Atovaquone Related Compound A (USP), Atovaquone Imp. B (EP), 296C85, USP Atovaquone Related Compound A, GR 261266X
IUPAC Name 2-[4-(4-chlorophenyl)cyclohexyl]-3-hydroxynaphthalene-1,4-dione
SMILES OC1=C([C@@H]2CC[C@@H](CC2)c3ccc(Cl)cc3)C(=O)c4ccccc4C1=O

Specifications

Analyte or Component Names cis-Atovaquone
Shipping Condition Room Temperature
InChI Formula InChI=1S/C22H19ClO3/c23-16-11-9-14(10-12-16)13-5-7-15(8-6-13)19-20(24)17-3-1-2-4-18(17)21(25)22(19)26/h1-4,9-13,15,26H,5-8H2/t13-,15+
Solution Type Neat
Industry Type Pharmaceutical
Quantity 10 mg
Formula Weight 366.1023
Chemical Name or Material cis-Atovaquone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.