CAS RN 137732-39-9
CAS RN 137732-39-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
cis-Atovaquone (Racemic), TRC
CAS: 137732-39-9 Molecular Formula: C22 H19 Cl O3 Molecular Weight (g/mol): 366.84 Synonym: cis-2-[4-(4-chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthoquinone,Atovaquone Related Compound A (USP),Atovaquone Imp. B (EP),296C85,USP Atovaquone Related Compound A,GR 261266X IUPAC Name: 2-[4-(4-chlorophenyl)cyclohexyl]-3-hydroxynaphthalene-1,4-dione SMILES: OC1=C([C@@H]2CC[C@@H](CC2)c3ccc(Cl)cc3)C(=O)c4ccccc4C1=O