missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHREBP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 116 publications
308.00€ - 625.00€
Specifications
| Dilution | Western Blot 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 21282101), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 26181104), Gel Super Shift Assays reported in scientific literature (PMID 20025850), SDS-Page reported in scientific literature (PMID 35041621), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31668386) |
|---|---|
| Gene Symbols | MLXIPL |
| Isotype | IgG |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18767263
|
Novus Biologicals
NB400-135SS |
308.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18767613
|
Novus Biologicals
NB400-135 |
625.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CHREBP Polyclonal specifically detects CHREBP in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays, Knockout Validated.Specifications
| Western Blot 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 21282101), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 26181104), Gel Super Shift Assays reported in scientific literature (PMID 20025850), SDS-Page reported in scientific literature (PMID 35041621), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31668386) | |
| MLXIPL | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title