Learn More
CHREBP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 116 publications
Brand:  Novus Biologicals NB400-135
 View more versions of this product
Product Code. 18767613
  • 625.00€ / 0.10mL
Description
CHREBP Polyclonal specifically detects CHREBP in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays, Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14carbohydrate response element binding protein
Affinity Purified
Western Blot 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 21282101), Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunoblotting reported in scientific literature (PMID 26181104), Gel Super Shift Assays reported in scientific literature (PMID 20025850), SDS-Page reported in scientific literature (PMID 35041621), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31668386)
MLXIPL
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.