missing translation for 'onlineSavingsMsg'
Learn More

Chikungunya Virus Antibody (8A2c), Novus Biologicals™

Mouse Monoclonal Antibody

203.00€ - 467.00€

Specifications

Antigen Chikungunya virus
Clone 8A2c
Concentration 1.0 mg/mL
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Functional
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Functional Assay
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18293890
View Documents
Novus Biologicals
NBP2-53117
0.1 mg
467.00€
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18675768
View Documents
Novus Biologicals
NBP2-53117-0.025mg
0.025 mg
203.00€
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chikungunya Virus Monoclonal specifically detects Chikungunya Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Chikungunya virus
1.0 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Functional Assay
Unconjugated
Mouse
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
PBS with 0.02% Sodium Azide
Live chikungunya virus
Primary
8A2c
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Functional
Monoclonal
Purified
RUO
Virus
Chikungunya
IgM
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.