missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chikungunya Virus Antibody (8A2c), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53117-0.025mg
This item is not returnable.
View return policy
Description
Chikungunya Virus Monoclonal specifically detects Chikungunya Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Chikungunya virus | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Functional | |
| Chikungunya | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgM |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| 8A2c | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| TMCO3 | |
| Live chikungunya virus | |
| 0.025 mg | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 55002 | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction