missing translation for 'onlineSavingsMsg'
Learn More

Chicken anti-Rat IgG whole molecule, FITC, Polyclonal, Secondary Antibody, Abnova™

Product Code. 16203140
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16203140 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16203140 Supplier Abnova Supplier No. PAB10845.1mg
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Antigen Chicken anti-Rat IgG whole molecule
Applications Flow Cytometry, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Description Chicken anti-rat IgG recognizes rat IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM potassium phosphate, 150mM NaCl, pH 7.2 (0.01% (w/v) sodium azide, 10mg/mL BSA (Immunoglobulin and Protease free))
Host Species Chicken
Immunogen Rat IgG whole molecule
Purification Method This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Quantity 1 mg
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.