missing translation for 'onlineSavingsMsg'
Learn More

Chicken anti-Rat IgG (H+L) Secondary Antibody, DyLight 405, Novus Biologicals™

Product Code. 18414518 Shop All Bio Techne Products
Modifier l'affichage
Click to view available options
Quantity:
1 mL
Conditionnement:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
18414518 1 mL 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18414518 Fournisseur Novus Biologicals Code fournisseur NBP175363V
 Restricted Item
Estimated Shipment: 16-10-2026
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Chicken Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot : 1:100-1:2000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Immunodiffusion, Immunoassay
Formulation 50mM Sodium Borate
Host Species Chicken
Immunogen This Chicken anti-Rat IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified rat IgG (H&L).
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.