missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Rat IgG (H+L) Secondary Antibody, Allophycocyanin/Cy7, R&D Systems™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion |
| Classification | Polyclonal |
| Conjugate | APC-Cyanine7 |
| Formulation | PBS |
| Host Species | Chicken |
| Immunogen | This Chicken anti-Rat IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified rat IgG (H&L). |
| Purification Method | Affinity purified |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?