missing translation for 'onlineSavingsMsg'
Learn More

Chicken anti-Rabbit IgG (H+L) Secondary Antibody, PE/Cy5.5, R&D Systems™

Product Code. 18575372 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18575372 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18575372 Supplier Novus Biologicals Supplier No. NB1206831PECY55
  • ONLINE EXCLUSIVE PRICE
    822.00€
    777.00€ / 500µL
    Save 45.00€
    5% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Chicken Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Western Blot, ELISA, Immunohistochemistry, Dot Blot, ChIP Assay
Classification Polyclonal
Conjugate PE-Cyanine5.5
Formulation PBS
Host Species Chicken
Immunogen Rabbit IgG whole molecule
Purification Method Multi-step
Quantity 500 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.