missing translation for 'onlineSavingsMsg'
Learn More

Chicken anti-Rabbit IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™

Product Code. 18523692 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18523692 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18523692 Supplier Novus Biologicals Supplier No. NBP173543
  • ONLINE EXCLUSIVE PRICE
    446.00€
    421.05€ / 1mg
    Save 24.95€
    5% Off
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chicken Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:2000 - 1:10000, ELISA 1:140000, Immunohistochemistry 1:1000 - 1:5000
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Host Species Chicken
Immunogen Chicken anti-Rabbit IgG (H+L) Secondary Antibody [HRP] was produced by repeated immunization with Rabbit IgG whole molecule in chicken.
Purification Method Immunogen affinity purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Reconstitution Reconstitute with 1.0 mL deionized water (or equivalent).
Target Species Rabbit
Content And Storage Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.