missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Rabbit IgG (H+L) Secondary Antibody, DyLight 755, R&D Systems™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Dilution | ELISA : 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay |
| Formulation | 50mM Sodium Borate |
| Host Species | Chicken |
| Immunogen | This Chicken anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified rabbit IgG (H&L). |
| Purification Method | Affinity purified |
| Quantity | 1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?