missing translation for 'onlineSavingsMsg'
Learn More

Chicken anti-Rabbit IgG (H+L) Secondary Antibody, DyLight 350, Novus Biologicals™

Product Code. 18425557 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425557 1 mL 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425557 Supplier Novus Biologicals Supplier No. NBP175265UV
  • ONLINE EXCLUSIVE PRICE
    412.00€
    389.55€ / 1mg
    Save 22.45€
    5% Off
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chicken Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay
Classification Polyclonal
Conjugate DyLight 350
Dilution ELISA : 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay
Formulation 50mM Sodium Borate
Host Species Chicken
Immunogen This Chicken anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified rabbit IgG (H&L).
Purification Method Affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.