missing translation for 'onlineSavingsMsg'
Learn More

Chicken anti-Rabbit IgG (H+L) Secondary Antibody, Biotin (Pre-adsorbed), Novus Biologicals™

Product Code. 18231022 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18231022 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18231022 Supplier Novus Biologicals Supplier No. NBP175267
  • ONLINE EXCLUSIVE PRICE
    250.00€
    236.25€ / 0.5mg
    Save 13.75€
    5% Off
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chicken Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Biotin
Dilution Western Blot 1:2000 - 1:20000, ELISA 1:2000 - 1:20000, Immunohistochemistry 1:500 - 1:5000, Immunohistochemistry-Paraffin
Host Species Chicken
Immunogen Purified Rabbit IgG (H&L)
Purification Method Affinity purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Based on IEP, this antibody reacts with heavy gamma chains on rabbit IgG and light chains on all rabbit immunoglobulins. This antibody has been pre-adsorbed against human and mouse IgG.
Target Species Rabbit
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.