missing translation for 'onlineSavingsMsg'
Learn More

Chicken anti-Mouse IgG (H+L) Secondary Antibody, PerCP, Novus Biologicals™

Product Code. 30232345 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30232345 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30232345 Supplier Novus Biologicals Supplier No. NBP175098PCP
  • ONLINE EXCLUSIVE PRICE
    322.00€
    304.50€ / 0.5mL
    Save 17.50€
    5% Off
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chicken Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Chemiluminescence Immunoassay, ELISA, Flow Cytometry, Immunoassay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunomicroscopy, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Chemiluminescence Immunoassay (CIA), ELISA, Flow Cytometry, Immunoassay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunomicroscopy, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Host Species Chicken
Immunogen This Chicken anti-Mouse IgG (H+L) Secondary Antibody was developed against purified mouse IgG (H&L).
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.