missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Mouse IgG (H+L) Secondary Antibody, DyLight 680, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Dilution | Western Blot : 1:100-1:2000, Flow Cytometry : 1:10-1:1000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence : 1:10 - 1:500, Immunoassay, Immunomicroscopy : 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA) |
| Formulation | 50mM Sodium Borate |
| Host Species | Chicken |
| Immunogen | This Chicken anti-Mouse IgG (H+L) Secondary Antibody was developed against purified mouse IgG (H&L). |
| Purification Method | Affinity purified |
| Quantity | 0.5 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?