missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Goat IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Chicken Polyclonal Antibody
187.00€ - 294.00€
Specifications
| Antigen | IgG (H+L) |
|---|---|
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoassay, Immunomicroscopy 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18531591
|
Novus Biologicals
NBP1-74804-0.5mg |
0.5 mg |
187.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18567101
|
Novus Biologicals
NBP1-74804 |
1 mg |
294.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence ImmunoassaySpecifications
| IgG (H+L) | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Chicken | |
| Adaptive Immunity, Immunology | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoassay, Immunomicroscopy 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Goat | |
| This Chicken anti-Goat IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified goat IgG (H&L). | |
| Secondary | |
| Store at 4°C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title