missing translation for 'onlineSavingsMsg'
Learn More

Chicken anti-Goat IgG (H+L) Secondary Antibody, PE, Novus Biologicals™

Product Code. 30232604 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30232604 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30232604 Supplier Novus Biologicals Supplier No. NB71058357PE
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chicken Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Host Species Chicken
Immunogen Chickens were immunized with purified, non-denatured goat IgG (containing both heavy and light chains) emulsified in Freund's adjuvant.
Purification Method Immunogen affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Goat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.