missing translation for 'onlineSavingsMsg'
Learn More

Chicken anti-Goat IgG (H+L) Secondary Antibody, Novus Biologicals™

Product Code. 18201905 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18201905 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18201905 Supplier Novus Biologicals Supplier No. NB710583570.5ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS pH 7.2 with 0.02% sodium azide
Gene Symbols IGHG1
Host Species Chicken
Immunogen Goat IgG
Quantity 0.5 mL
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity There is no detectable cross-reactivity with rabbit, human or mouse IgG.
Target Species Goat
Content And Storage Store at 4°C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.