missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chicken anti-Goat IgG (H+L) Secondary Antibody, Allophycocyanin/Cy7, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB710-58357APCCY7
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:1000 | |
| Chicken | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgY |
| Western Blot | |
| APC-Cyanine7 | |
| PBS | |
| Chickens were immunized with purified, non-denatured goat IgG (containing both heavy and light chains) emulsified in Freund's adjuvant. | |
| 500 μg | |
| Adaptive Immunity, Immunology | |
| Goat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction