missing translation for 'onlineSavingsMsg'
Learn More

CEBP Beta Antibody (3H9), Novus Biologicals™

Mouse Monoclonal Antibody

203.00€ - 552.00€

Specifications

Antigen CEBP Beta
Clone 3H9
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18124288
View Documents
Novus Biologicals
NBP2-37567
0.1 mL
552.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18642476
View Documents
Novus Biologicals
NBP2-37567-0.025ml
0.025 mL
203.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CEBP Beta Monoclonal specifically detects CEBP Beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

CEBP Beta
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000
Monoclonal
Ascites
RUO
Ascites with 0.03% Sodium Azide
1051
Purified recombinant fragment of human CEBPB expressed in E. Coli.
Primary
36 kDa
3H9
Western Blot, Flow Cytometry, ELISA
Unconjugated
Mouse
Human
C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CCAAT/enhancer-binding protein beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAPTCF-5, Liver activator protein, liver-enriched transcriptional activator protein, MGC32080, NF-IL6, Nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5NFIL6, Transcription factor 5
CEBPB
IgG1
Unpurified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.