missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBP Beta Antibody (3H9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37567
This item is not returnable.
View return policy
Description
CEBP Beta Monoclonal specifically detects CEBP Beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| CEBP Beta | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| CEBPB | |
| Purified recombinant fragment of human CEBPB expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 1051 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA | |
| 3H9 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CCAAT/enhancer-binding protein beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAPTCF-5, Liver activator protein, liver-enriched transcriptional activator protein, MGC32080, NF-IL6, Nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5NFIL6, Transcription factor 5 | |
| Mouse | |
| 36 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction