missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Cdk7 Antibody (MO-1), Novus Biologicals™

Mouse Monoclonal Antibody

213.00€ - 508.00€

Specifications

Antigen Cdk7
Clone MO-1
Dilution Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30066840
View Documents
Novus Biologicals™
NBP3-30978-25ug
25 μg
213.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30066957
View Documents
Novus Biologicals™
NBP3-30978-100ug
100 μg
508.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cdk7 Monoclonal antibody specifically detects Cdk7 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications

Specifications

Cdk7
Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation
Monoclonal
Purified
RUO
Human
CAK1Xenopus laevis, cdk-activating kinase), CDK-activating kinase, Cell division protein kinase 7, cyclin-dependent kinase 7, cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, EC 2.7.11.24, p39 Mo15, serine/threonine kinase stk1,39 kDa protein kinase, serine/threonine protein kinase 1, serine/threonine protein kinase MO15, TFIIH basal transcription factor complex kinase subunit
Rabbits were immunized with a recombinant 221 amino acid fragment of the C terminus of Cdk7 recognizing a 40kD protein
Primary
Store at -20°C. Avoid freeze/thaw cycles.
MO-1
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Mouse
Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators
Phosphate buffered saline
1022
IgG2b
Ammonium sulfate precipitation
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.