Learn More
Novus Biologicals™ Cdk7 Antibody (MO-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30978-25ug
This item is not returnable.
View return policy
Description
Cdk7 Monoclonal antibody specifically detects Cdk7 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| Cdk7 | |
| Monoclonal | |
| Unconjugated | |
| Phosphate buffered saline | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| MO-1 | |
| Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation | |
| CAK1Xenopus laevis, cdk-activating kinase), CDK-activating kinase, Cell division protein kinase 7, cyclin-dependent kinase 7, cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, EC 2.7.11.24, p39 Mo15, serine/threonine kinase stk1,39 kDa protein kinase, serine/threonine protein kinase 1, serine/threonine protein kinase MO15, TFIIH basal transcription factor complex kinase subunit | |
| Rabbits were immunized with a recombinant 221 amino acid fragment of the C terminus of Cdk7 recognizing a 40kD protein | |
| 25 μg | |
| Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators | |
| 1022 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG2b |