missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdk5 Antibody (4E4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37602-0.025ml
This item is not returnable.
View return policy
Description
Cdk5 Monoclonal specifically detects Cdk5 in Human, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cdk5 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| CDK5 | |
| Purified recombinant fragment of human CDK5 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Rat, Monkey | |
| Ascites |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 40000 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit | |
| Mouse | |
| 35 kDa | |
| 0.025 mL | |
| Cell Cycle and Replication | |
| 1020 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction