missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdk5 Antibody (4E4), Novus Biologicals™
Mouse Monoclonal Antibody
196.00€ - 538.00€
Specifications
| Antigen | Cdk5 |
|---|---|
| Clone | 40000 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18135548
|
Novus Biologicals
NBP2-37602 |
0.1 mL |
538.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631976
|
Novus Biologicals
NBP2-37602-0.025ml |
0.025 mL |
196.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cdk5 Monoclonal specifically detects Cdk5 in Human, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Cdk5 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Rat, Monkey | |
| Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit | |
| CDK5 | |
| IgG1 | |
| Unpurified |
| 40000 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| Ascites with 0.03% Sodium Azide | |
| 1020 | |
| Purified recombinant fragment of human CDK5 expressed in E. Coli. | |
| Primary | |
| 35 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title