missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD69 Antibody (8B6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-51607-0.025ml
This item is not returnable.
View return policy
Description
CD69 Monoclonal specifically detects CD69 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD69 | |
| Monoclonal | |
| Unconjugated | |
| Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3 | |
| Mouse | |
| 22.5 kDa | |
| 0.025 mL | |
| Innate Immunity | |
| 969 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 8B6 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Imaging Mass Cytometry | |
| CD69 | |
| Purified recombinant fragment of human CD69 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction