missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD69 Antibody (8B6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
203.00€
Specifications
| Antigen | CD69 |
|---|---|
| Clone | 8B6 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Imaging Mass Cytometry |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
CD69 Monoclonal specifically detects CD69 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CD69 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Imaging Mass Cytometry | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 969 | |
| Purified recombinant fragment of human CD69 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 8B6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Innate Immunity | |
| Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3 | |
| CD69 | |
| IgG1 | |
| Unpurified | |
| 22.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title