missing translation for 'onlineSavingsMsg'
Learn More

CD46 Antibody (7D12), Novus Biologicals™

Rabbit Monoclonal Antibody

343.00€ - 533.00€

Specifications

Antigen CD46
Clone 7D12
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043831
View Documents
Novus Biologicals
NBP3-26679-50ul
50 μL
343.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043839
View Documents
Novus Biologicals
NBP3-26679-100ul
100 μL
533.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD46 Monoclonal antibody specifically detects CD46 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications

Specifications

CD46
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human
AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen
A synthesized peptide derived from Human CD46 [UniProt P15529]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
7D12
Western Blot, ELISA, Immunohistochemistry
Unconjugated
Rabbit
Cancer, Cell Biology, Cellular Markers, Immunology
PBS, pH 7.4, 150mM NaCl and 50% glycerol
4179
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.