Learn More
CD39/ENTPD1 Antibody (17B5F11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
226.00€ - 500.00€
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18705403
|
Novus Biologicals
NBP2-25223SS |
226.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18775733
|
Novus Biologicals
NBP2-25223 |
0.1 mg |
500.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD39/ENTPD1 Monoclonal specifically detects CD39/ENTPD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Human, Mouse |