Learn More
CD39/ENTPD1 Antibody (17B5F11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-25223
This item is not returnable.
View return policy
Description
CD39/ENTPD1 Monoclonal specifically detects CD39/ENTPD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD39/ENTPD1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 0.5 - 5 ug/mL, ELISA reported in scientific literature (PMID 33488292), Immunohistochemistry 1:20 - 1:1000, Immunohistochemistry-Paraffin 5 ug/mL | |
| ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Human, Mouse and Rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b λ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 17B5F11 | |
| Unconjugated | |
| NP_001191742 | |
| ENTPD1 | |
| Amino acids 102-208 of human CD39 were used as the immunogen for the antibody. | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 953 | |
| Human, Mouse, Rat | |
| Purified |