missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 (PD-1) Mouse anti-Human, BB700, Clone: EH12.1 (also known as EH12), BD Horizon™
Mouse Monoclonal Antibody
219.24€ - 492.39€
Specifications
| Antigen | CD279 (PD-1) |
|---|---|
| Clone | EH12.1 (also known as EH12) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 700 (BB700) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15885069
|
BD Biosciences
566461 |
25 Tests |
220.00€ 219.24€ / Each Save 0.76€ 0.35% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15875069
|
BD Biosciences
566460 |
100 Tests |
493.00€ 492.39€ / Each Save 0.61€ 0.12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow Cytometry.Specifications
| CD279 (PD-1) | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Human | |
| Primary |
| EH12.1 (also known as EH12) | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title