missing translation for 'onlineSavingsMsg'
Learn More

CD279 (PD-1) Mouse anti-Human, BB700, Clone: EH12.1 (also known as EH12), BD Horizon™

Mouse Monoclonal Antibody

Brand:  BD Biosciences 566461

 View more versions of this product

Product Code. 15885069

  • ONLINE EXCLUSIVE PRICE
    220.00€
    219.24€ / Each
    Save 0.76€
    0.35% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD279 (PD-1)
Monoclonal
Brilliant Blue 700 (BB700)
25 Tests
Primary
Refrigerate (2 to 8°C)
IgG1 κ
Flow Cytometry
EH12.1 (also known as EH12)
Mouse
RUO
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.