Learn More
CD206 Mouse anti-Human, APC, Clone: 19.2, BD
Mouse Monoclonal Antibody
74.90€ - 293.00€
Specifications
| Antigen | CD206 |
|---|---|
| Clone | 19.2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15881309
|
BD Biosciences
561763 |
25 Tests |
74.90€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15806428
|
BD Biosciences
550889 |
100 Tests |
293.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 19.2 monoclonal antibody specifically binds to CD206 which is also known as the macrophage mannose receptor (MMR) or C-type lectin domain family 13 member D (CLEC13D). CD206 is a type I transmembrane glycoprotein of approximately 162 kDa which binds to glycoconjugates containing mannose, fucose, or N-acetylglucosamine. These carbohydrates are present on the surface of many microorganisms and enable the macrophage to bind to microorganisms through the MMR and internalize them during the process of phagocytosis. CD206 is expressed on human macrophages, endothelial cells, and cultured dendritic cells. It is not detected on resting monocytes. Mannose receptor expression is upregulated on peripheral blood mononuclear cells following 3 day incubation with GM-CSF.
Specifications
| CD206 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| Macrophage mannose receptor |
| 19.2 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human Placental Macrophage Mannose Receptor | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.