missing translation for 'onlineSavingsMsg'
Learn More

CD206 Mouse anti-Human, APC, Clone: 19.2, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 550889

 View more versions of this product

Product Code. 15806428

  • ONLINE EXCLUSIVE PRICE
    324.00€
    323.62€ / Each
    Save 0.38€
    0.12% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 19.2 monoclonal antibody specifically binds to CD206 which is also known as the macrophage mannose receptor (MMR) or C-type lectin domain family 13 member D (CLEC13D). CD206 is a type I transmembrane glycoprotein of approximately 162 kDa which binds to glycoconjugates containing mannose, fucose, or N-acetylglucosamine. These carbohydrates are present on the surface of many microorganisms and enable the macrophage to bind to microorganisms through the MMR and internalize them during the process of phagocytosis. CD206 is expressed on human macrophages, endothelial cells, and cultured dendritic cells. It is not detected on resting monocytes. Mannose receptor expression is upregulated on peripheral blood mononuclear cells following 3 day incubation with GM-CSF.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD206
Monoclonal
APC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human Placental Macrophage Mannose Receptor
100 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
19.2
Macrophage mannose receptor
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.