missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Rat, anti-Mouse, Clone: 1D3, Proteintech
Rat Monoclonal Antibody
50.00€ - 149.00€
Specifications
| Antigen | CD19 |
|---|---|
| Clone | 1D3 |
| Concentration | 0.5 mg/ml |
| Applications | Flow Cytometry |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18066395
|
Proteintech
65290-1-IG-500UG |
500 μg |
149.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18004285
|
Proteintech
65290-1-IG-100UG |
100 μg |
50.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD19 antibody from Proteintech is a rat monoclonal antibody generated with mouse cd19 transfected cell line of mouse CD19. This antibody recognizes mouse antigen. The CD19 antibody has been validated for the following applications: FC analysis.
Specifications
| CD19 | |
| 0.5 mg/ml | |
| Liquid | |
| Mouse | |
| AW495831, Cd19, CD19 antigen | |
| Primary |
| 1D3 | |
| Flow Cytometry | |
| Rat | |
| 12478 | |
| IgG2a κ | |
| Store at 2°C to 8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title