missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Rat, anti-Mouse, Clone: 1D3, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65290-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD19 antibody from Proteintech is a rat monoclonal antibody generated with mouse cd19 transfected cell line of mouse CD19. This antibody recognizes mouse antigen. The CD19 antibody has been validated for the following applications: FC analysis.
Specifications
| CD19 | |
| 1D3 | |
| AW495831, Cd19, CD19 antigen | |
| 500 μg | |
| 12478 | |
| Store at 2°C to 8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| 0.5 mg/ml | |
| Rat | |
| Primary | |
| Mouse | |
| 150 kDa | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction