missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Antibody (C19/366), Novus Biologicals™
Mouse Monoclonal Antibody
277.00€ - 516.00€
Specifications
| Antigen | CD19 |
|---|---|
| Clone | C19/366 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 0.5-1.0ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18465971
|
Novus Biologicals
NBP2-29424-20ug |
20 ug |
277.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18173113
|
Novus Biologicals
NBP2-29424 |
0.1 mg |
516.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CD19 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| P15391 | |
| 930 | |
| Recombinant full-length human CD19 protein (Uniprot: P15391) | |
| Primary | |
| Store at 4C. |
| C19/366 | |
| Immunohistochemistry-Paraffin 0.5-1.0ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| CD19 | |
| IgG1 κ | |
| Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title