missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Antibody (C19/366), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29424-20ug
This item is not returnable.
View return policy
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD19 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 0.5-1.0ug/ml | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| C19/366 | |
| Unconjugated | |
| P15391 | |
| CD19 | |
| Recombinant full-length human CD19 protein (Uniprot: P15391) | |
| 20 ug | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 930 | |
| Store at 4C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction