missing translation for 'onlineSavingsMsg'
Learn More

CD133 Mouse anti-Human, FITC, Clone: W6B3C1 (also known as W6B3), BD Biosciences™

Mouse Monoclonal Antibody

128.50€ - 343.38€

Specifications

Antigen CD133
Clone W6B3C1 (also known as W6B3)
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17277814
View Documents
BD Biosciences
567033
25 μg
ONLINE EXCLUSIVE PRICE
129.00€
128.50€ / 25µg
Save 0.50€
0.39% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17237814
View Documents
BD Biosciences
567029
0.1 mg
ONLINE EXCLUSIVE PRICE
344.00€
343.38€ / 100µg
Save 0.62€
0.18% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD133 Monoclonal antibody specifically detects CD133 in Human samples. It is validated for Flow cytometry
Specifications

Specifications

CD133
0.5 mg/mL
Monoclonal
Mouse
Human
PROM1; PROML1; prominin-1; AC133; CORD12; MCDR2; MSTP061; RP41; STGD4
IgG1 κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with FITC under optimum conditions, and unreacted FITC was removed.
W6B3C1 (also known as W6B3)
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
WERI-RB-1 Retinoblastoma Cell
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.