missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133 Mouse anti-Human, FITC, Clone: W6B3C1 (also known as W6B3), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 567033
This item is not returnable.
View return policy
Description
CD133 Monoclonal antibody specifically detects CD133 in Human samples. It is validated for Flow cytometry
Specifications
| CD133 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with FITC under optimum conditions, and unreacted FITC was removed. |
| Flow Cytometry | |
| W6B3C1 (also known as W6B3) | |
| FITC | |
| PROM1; PROML1; prominin-1; AC133; CORD12; MCDR2; MSTP061; RP41; STGD4 | |
| WERI-RB-1 Retinoblastoma Cell | |
| RUO | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction